siRNA Details
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191789.1 | NP_001178718.1 | 27 | 3706 | ||
XM_006242190.2 | XP_006242252.1 | 27 | 4101 | |||
XM_008765689.1 | XP_008763911.1 | 27 | 4101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
