siRNA Details
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100887.1 | NP_001094357.1 | 8 | 1224 | ||
XM_006242192.2 | XP_006242254.1 | 7 | 1132 | |||
GenBank mRNA | AY623036.1 | - | 6 | 827 | ||
BC167068.1 | - | 7 | 978 | 8363586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
