siRNA Details
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100887.1 | NP_001094357.1 | 3 | 501 | ||
XM_006242192.2 | XP_006242254.1 | 2 | 409 | |||
GenBank mRNA | AY623036.1 | - | 1 | 104 | ||
BC167068.1 | - | 2 | 255 | 8363586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
