siRNA Details
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013105.2 | NP_001013123.2 | 3 | 315 | ||
XM_006257344.2 | XP_006257406.1 | 2 | 1220 | |||
XM_008765695.1 | XP_008763917.1 | 2 | 218 | |||
XM_008765696.1 | XP_008763918.1 | 2 | 230 | |||
XM_008765697.1 | XP_008763919.1 | 2 | 207 | |||
GenBank mRNA | BC084709.1 | - | 3 | 293 | 7301886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
