siRNA Details
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191615.2 | NP_001178544.1 | 3 | 551 | ||
XM_006257354.2 | XP_006257416.1 | 4 | 1016 | |||
XM_006257355.2 | XP_006257417.1 | 4 | 699 |
