siRNA Details
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013107.1 | NP_001013125.1 | 4 | 456 | ||
GenBank mRNA | BC062052.1 | - | 4 | 456 | 5598966 |

More Information
Other Species Targeted

Gene Ontology Categories:
