siRNA Details
Gene Symbol: Myg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005545.1 | NP_001005545.1 | 6 | 828 | ||
GenBank mRNA | BC082112.1 | - | 6 | 828 | 7135321 | |
FQ213994.1 | - | 4 | 893 | |||
FQ230067.1 | - | 6 | 858 |
