siRNA Details
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106795.1 | NP_001100265.1 | 12 | 1262 | ||
XM_006242387.2 | XP_006242449.1 | 11 | 1179 | |||
XM_008765704.1 | XP_008763926.1 | 11 | 1222 | |||
XM_008765705.1 | XP_008763927.1 | 11 | 1203 |
