siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013951.1 | NP_001013973.1 | 3 | 307 | ||
GenBank mRNA | BC088233.1 | - | 3 | 307 | 7324702 | |
FQ216283.1 | - | 3 | 319 | |||
FQ229089.1 | - | 3 | 333 |
Gene Symbol: LOC103691612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761229.1 | XP_008759451.1 | 1 | 1599 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
