siRNA Details
Gene Symbol: Mmp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106800.1 | NP_001100270.1 | 10 | 1411 | ||
XM_006242487.2 | XP_006242549.1 | 11 | 1680 |

More Information
Other Species Targeted

Gene Ontology Categories:
