siRNA Details
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106801.1 | NP_001100271.1 | 4 | 821 | ||
GenBank mRNA | BC083809.1 | - | 4 | 721 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106801.1 | NP_001100271.1 | 4 | 821 | ||
GenBank mRNA | BC083809.1 | - | 4 | 721 |