siRNA Details
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106802.1 | NP_001100272.1 | 12, 13 | 1436 | ||
XM_006242562.2 | XP_006242624.1 | 12, 13 | 1784 | |||
XM_006242568.2 | XP_006242630.1 | 11, 12 | 1650 | |||
XM_008765922.1 | XP_008764144.1 | 9, 10 | 1293 | |||
XM_008765923.1 | XP_008764145.1 | 8, 9 | 1194 | |||
XM_008765924.1 | XP_008764146.1 | 10, 11 | 1404 | |||
XM_008765925.1 | XP_008764147.1 | 12, 13 | 1301 | |||
XM_008765926.1 | XP_008764148.1 | 11, 12 | 1637 | |||
XM_008765927.1 | XP_008764149.1 | 3, 4 | 242 |
