siRNA Details
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106806.2 | NP_001100276.1 | 14, 15 | 1338 | ||
XM_006242633.2 | XP_006242695.1 | 14, 15 | 1406 | |||
XM_006242634.1 | XP_006242696.1 | 14, 15 | 1471 | |||
GenBank mRNA | BC161987.1 | - | 14, 15 | 1338 | 9030870 |
