siRNA Details
Gene Symbol: Bmper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135799.1 | NP_001129271.1 | 14 | 2321 | ||
XM_006242715.2 | XP_006242777.1 | 12 | 3275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
