siRNA Details
Gene Symbol: Olr1304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000464.1 | NP_001000464.1 | 1 | 275 |
Gene Symbol: Olr1305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000959.1 | NP_001000959.1 | 1 | 275 |
