siRNA Details
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106816.1 | NP_001100286.1 | 3 | 663 | ||
XM_006242915.2 | XP_006242977.1 | 3 | 6395 | |||
XM_006242916.2 | XP_006242978.1 | 3 | 753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
