siRNA Details
Gene Symbol: C2cd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011996.1 | NP_001011996.1 | 6 | 1215 | ||
XM_006242917.2 | XP_006242979.1 | 6 | 1266 | |||
XM_006242918.2 | XP_006242980.1 | 6 | 1266 | |||
XM_006242919.2 | XP_006242981.1 | 6 | 1266 | |||
XM_006242920.2 | XP_006242982.1 | 6 | 1266 | |||
XM_008766156.1 | XP_008764378.1 | 6 | 1266 | |||
XM_008766157.1 | XP_008764379.1 | 6 | 818 | |||
GenBank mRNA | BC085925.1 | - | 6 | 1215 | 7109537 |
