siRNA Details
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212516.2 | NP_997681.1 | 2 | 215 | ||
GenBank mRNA | BC058497.1 | - | 2 | 216 | 6918545 | |
FQ210248.1 | - | 2 | 205 | |||
FQ211867.1 | - | 2 | 212 | |||
FQ214576.1 | - | 211 | ||||
FQ217032.1 | - | 2 | 198 | |||
FQ217139.1 | - | 2 | 231 | |||
FQ217410.1 | - | 2 | 232 | |||
FQ217906.1 | - | 2 | 204 | |||
FQ218008.1 | - | 2 | 204 | |||
FQ221909.1 | - | 2 | 244 | |||
FQ223878.1 | - | 2 | 205 | |||
FQ223920.1 | - | 2 | 211 | |||
FQ224004.1 | - | 2 | 205 | |||
FQ224067.1 | - | 2 | 205 | |||
FQ224620.1 | - | 2 | 205 | |||
FQ229120.1 | - | 2 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
