siRNA Details
Gene Symbol: Ttc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004246.2 | NP_001004246.1 | 10 | 882 | ||
XM_006243004.2 | XP_006243066.1 | 11 | 1695 | |||
XM_006243005.2 | XP_006243067.1 | 11 | 950 | |||
XM_008766196.1 | XP_008764418.1 | 11 | 1097 | |||
GenBank mRNA | BC079409.1 | - | 10 | 884 | 7113087 |
