siRNA Details
Gene Symbol: Stoml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243184.2 | XP_006243246.1 | 2 | 288 | ||
XM_236297.8 | XP_236297.3 | 2 | 288 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stoml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243184.2 | XP_006243246.1 | 2 | 288 | ||
XM_236297.8 | XP_236297.3 | 2 | 288 |