siRNA Details
Gene Symbol: Rec114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106825.1 | NP_001100295.1 | 3 | 363 | ||
XM_006243157.2 | XP_006243219.1 | 3 | 495 | |||
XM_006243158.2 | XP_006243220.1 | 3 | 508 | |||
XM_006243159.2 | XP_006243221.1 | 3 | 503 | |||
XM_006243160.2 | XP_006243222.1 | 3 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
