siRNA Details
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106829.2 | NP_001100299.2 | 6 | 811 | ||
GenBank mRNA | AY644724.1 | - | 6 | 811 |

More Information
Other Species Targeted

Gene Ontology Categories:
