siRNA Details
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106829.2 | NP_001100299.2 | 7 | 941 | ||
GenBank mRNA | AY644724.1 | - | 7 | 941 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019663, AK075708, AF077950, AK153871, AK035682, and others.
Gene Ontology Categories:
