siRNA Details
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006987.1 | NP_001006988.1 | 7 | 814 | ||
XM_006243251.2 | XP_006243313.1 | 7 | 845 | |||
GenBank mRNA | BC083760.1 | - | 7 | 814 | 7192933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
