siRNA Details
Gene Symbol: Ns5atp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201418.1 | NP_958821.1 | 2, 3 | 121 | ||
GenBank mRNA | AY496944.1 | - | 2, 3 | 121 | ||
FQ220183.1 | - | 2, 3 | 198 |

More Information
Other Species Targeted

Gene Ontology Categories:
