siRNA Details
Gene Symbol: Snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106832.1 | NP_001100302.1 | 4 | 362 | ||
XM_006243304.2 | XP_006243366.1 | 4 | 414 | |||
XM_006243305.2 | XP_006243367.1 | 4 | 437 | |||
XM_008766240.1 | XP_008764462.1 | 4 | 438 |

More Information
Other Species Targeted

Gene Ontology Categories:
