siRNA Details
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106833.1 | NP_001100303.1 | 6 | 1468 | ||
XM_008766242.1 | XP_008764464.1 | 5 | 1285 | |||
GenBank mRNA | FQ230750.1 | - | 3 | 457 |
