siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106836.1 | NP_001100306.1 | 10 | 2680 | ||
XM_006243372.2 | XP_006243434.1 | 10 | 2833 | |||
XM_006243373.2 | XP_006243435.1 | 10 | 2832 | |||
XM_006243374.2 | XP_006243436.1 | 10 | 2829 | |||
XM_006243376.2 | XP_006243438.1 | 11 | 2925 | |||
XM_006243377.2 | XP_006243439.1 | 10 | 2862 | |||
XM_008766243.1 | XP_008764465.1 | 10 | 2829 | |||
XM_008766244.1 | XP_008764466.1 | 10 | 2611 | |||
XM_008766245.1 | XP_008764467.1 | 10 | 2860 |
