siRNA Details
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005549.1 | NP_001005549.1 | 10 | 1405 | ||
GenBank mRNA | BC081887.1 | - | 10 | 1405 | 7109073 |

More Information
Other Species Targeted

Gene Ontology Categories:
