siRNA Details
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005549.1 | NP_001005549.1 | 7 | 992 | ||
GenBank mRNA | BC081887.1 | - | 7 | 992 | 7109073 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005549.1 | NP_001005549.1 | 7 | 992 | ||
GenBank mRNA | BC081887.1 | - | 7 | 992 | 7109073 |