siRNA Details
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106842.2 | NP_001100312.2 | 4 | 532 | ||
XM_006243436.1 | XP_006243498.1 | 4 | 908 | |||
XM_006243437.1 | XP_006243499.1 | 2 | 191 | |||
GenBank mRNA | FQ213152.1 | - | 1 | 105 | ||
FQ225293.1 | - | 4 | 722 |
