siRNA Details
Gene Symbol: Trim43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142944.1 | NP_001136416.1 | 6 | 1151 | ||
XM_006243512.2 | XP_006243574.1 | 7 | 1391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
