siRNA Details
Gene Symbol: Plscr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012000.1 | NP_001012000.1 | 6 | 800 | ||
GenBank mRNA | AY548832.1 | - | 6 | 800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
