siRNA Details
Gene Symbol: Cldn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106846.2 | NP_001100316.1 | 2 | 543 | ||
XM_006234262.2 | XP_006234324.1 | 2 | 646 | |||
GenBank mRNA | CB790981.1 | - | 1 | 320 |
