siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106847.2 | NP_001100317.2 | 12 | 1387 | ||
XM_006243556.2 | XP_006243618.1 | 1691 | ||||
XM_006243558.2 | XP_006243620.1 | 12 | 2790 | |||
XM_006243561.2 | XP_006243623.1 | 11 | 2750 | |||
XM_006243563.1 | XP_006243625.1 | 14 | 1799 | |||
XM_006243566.2 | XP_006243628.1 | 1992 | ||||
XM_006243567.2 | XP_006243629.1 | 13 | 3114 | |||
XM_006243568.2 | XP_006243630.1 | 14 | 3211 | |||
XM_006243569.2 | XP_006243631.1 | 13 | 1925 | |||
XM_006243571.2 | XP_006243633.1 | 13 | 1878 | |||
XM_006243572.2 | XP_006243634.1 | 11 | 2746 | |||
XM_008766479.1 | XP_008764701.1 | 13 | 1369 | |||
XM_008766480.1 | XP_008764702.1 | 13 | 1365 | |||
XM_008766481.1 | XP_008764703.1 | 13 | 1369 | |||
XM_008766482.1 | XP_008764704.1 | 17 | 1906 | |||
XM_008766483.1 | XP_008764705.1 | 13 | 1692 | |||
XM_008766484.1 | XP_008764706.1 | 17 | 1760 | |||
XM_008766485.1 | XP_008764707.1 | 16 | 1664 | |||
XM_008766487.1 | XP_008764709.1 | 14 | 1495 | |||
XM_008766488.1 | XP_008764710.1 | 11 | 1205 | |||
GenBank mRNA | BC168961.1 | - | 12 | 1387 | 7444676 |
