siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106847.2 | NP_001100317.2 | 11 | 1224 | ||
XM_006243556.2 | XP_006243618.1 | 1528 | ||||
XM_006243558.2 | XP_006243620.1 | 11 | 2627 | |||
XM_006243561.2 | XP_006243623.1 | 10 | 2587 | |||
XM_006243563.1 | XP_006243625.1 | 13 | 1636 | |||
XM_006243566.2 | XP_006243628.1 | 1829 | ||||
XM_006243567.2 | XP_006243629.1 | 12 | 2951 | |||
XM_006243568.2 | XP_006243630.1 | 13 | 3048 | |||
XM_006243569.2 | XP_006243631.1 | 12 | 1762 | |||
XM_006243571.2 | XP_006243633.1 | 12 | 1715 | |||
XM_006243572.2 | XP_006243634.1 | 10 | 2583 | |||
XM_008766479.1 | XP_008764701.1 | 12 | 1206 | |||
XM_008766480.1 | XP_008764702.1 | 12 | 1202 | |||
XM_008766481.1 | XP_008764703.1 | 12 | 1206 | |||
XM_008766482.1 | XP_008764704.1 | 16 | 1743 | |||
XM_008766483.1 | XP_008764705.1 | 12 | 1529 | |||
XM_008766484.1 | XP_008764706.1 | 16 | 1597 | |||
XM_008766485.1 | XP_008764707.1 | 15 | 1501 | |||
XM_008766487.1 | XP_008764709.1 | 13 | 1332 | |||
XM_008766488.1 | XP_008764710.1 | 10 | 1042 | |||
GenBank mRNA | BC168961.1 | - | 11 | 1224 | 7444676 |

More Information
Other Species Targeted

Gene Ontology Categories:
