siRNA Details
Gene Symbol: Rnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106848.1 | NP_001100318.1 | 2 | 234 | ||
XM_008766489.1 | XP_008764711.1 | 2 | 593 |

More Information
Other Species Targeted

Gene Ontology Categories:
