siRNA Details
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037772.1 | NP_001032861.2 | 11 | 1359 | ||
NM_001244805.1 | NP_001231734.1 | 11 | 1528 | |||
XM_008766490.1 | XP_008764712.1 | 13 | 1650 | |||
XM_008766491.1 | XP_008764713.1 | 12 | 1374 | |||
XM_008766492.1 | XP_008764714.1 | 11 | 2949 | |||
XM_008766493.1 | XP_008764715.1 | 13 | 1525 | |||
GenBank mRNA | BC083632.1 | - | 14 | 1612 | ||
BC098948.1 | - | 11 | 1359 | |||
BC161815.1 | - | 11 | 1528 | 9031816 |
