siRNA Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013252.1 | NP_001013270.1 | 4 | 359 | ||
GenBank mRNA | AY325214.1 | - | 2507 | |||
AY325230.1 | - | 2510 | ||||
AY325241.1 | - | 2507 | ||||
BC098951.1 | - | 351 | 7324816 | |||
BF523222.1 | - | 1 | 25 | |||
BF525252.1 | - | 365 |
