siRNA Details
Gene Symbol: RGD1307461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106854.1 | NP_001100324.1 | 3 | 599 | ||
XM_006243731.2 | XP_006243793.1 | 3 | 1443 | |||
XM_006243732.2 | XP_006243794.1 | 2 | 1323 | |||
XM_006243733.2 | XP_006243795.1 | 2 | 1317 | |||
XM_008766503.1 | XP_008764725.1 | 2 | 2240 | |||
GenBank mRNA | FQ224195.1 | - | 3 | 612 |
