siRNA Details
Gene Symbol: Hyal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207599.2 | NP_997482.2 | 4 | 1239 | ||
XM_008766504.1 | XP_008764726.1 | 3 | 1148 | |||
GenBank mRNA | AB100602.1 | - | 3 | 1103 | ||
BC097259.1 | - | 4 | 1239 | 7453545 |
