siRNA Details
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199099.2 | NP_954530.1 | 2, 3 | 207 | ||
XM_006243751.2 | XP_006243813.1 | 2, 3 | 248 | |||
GenBank mRNA | BC060585.1 | - | 2, 3 | 208 | 6891123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
