siRNA Details
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004250.2 | NP_001004250.1 | 3 | 279 | ||
GenBank mRNA | BC078923.1 | - | 3 | 281 | 7106265 | |
FQ213124.1 | - | 3 | 286 | |||
FQ216746.1 | - | 3 | 282 | |||
FQ224157.1 | - | 3 | 287 | |||
FQ229052.1 | - | 3 | 302 | |||
FQ231552.1 | - | 3 | 301 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
