siRNA Details
Gene Symbol: Shisa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006989.1 | NP_001006990.1 | 3 | 245 | ||
XM_006243760.1 | XP_006243822.1 | 5 | 577 | |||
XM_006243761.1 | XP_006243823.1 | 5 | 574 | |||
XM_006243762.2 | XP_006243824.1 | 3 | 563 | |||
XM_008766511.1 | XP_008764733.1 | 4 | 275 | |||
GenBank mRNA | BC083710.1 | - | 3 | 245 | 7190608 | |
FQ214071.1 | - | 3 | 221 | |||
FQ227338.1 | - | 3 | 250 | |||
FQ231597.1 | - | 5 | 561 |
