siRNA Details
Gene Symbol: Lrrfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024761.1 | NP_001019932.1 | 9 | 937 | ||
XM_006243939.2 | XP_006244001.1 | 21 | 1722 | |||
XM_006243957.1 | XP_006244019.1 | 8 | 976 | |||
XM_006243958.2 | XP_006244020.1 | 8 | 916 | |||
XM_006243960.1 | XP_006244022.1 | 7 | 904 | |||
XM_008766616.1 | XP_008764838.1 | 21 | 1723 | |||
XM_008766617.1 | XP_008764839.1 | 21 | 1486 | |||
XM_008766618.1 | XP_008764840.1 | 21 | 1699 | |||
XM_008766619.1 | XP_008764841.1 | 20 | 1652 | |||
XM_008766620.1 | XP_008764842.1 | 20 | 1678 | |||
XM_008766621.1 | XP_008764843.1 | 20 | 1672 | |||
XM_008766622.1 | XP_008764844.1 | 20 | 1654 | |||
XM_008766623.1 | XP_008764845.1 | 20 | 1653 | |||
XM_008766624.1 | XP_008764846.1 | 20 | 1633 | |||
XM_008766625.1 | XP_008764847.1 | 21 | 1723 | |||
XM_008766626.1 | XP_008764848.1 | 19 | 1603 | |||
XM_008766627.1 | XP_008764849.1 | 19 | 1563 | |||
XM_008766628.1 | XP_008764850.1 | 18 | 1557 | |||
XM_008766629.1 | XP_008764851.1 | 17 | 1488 | |||
GenBank mRNA | BC097955.1 | - | 9 | 937 | 7456701 | |
FQ233539.1 | - | 8 | 960 |
