siRNA Details
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191819.1 | NP_001178748.1 | 3 | 493 | ||
XM_006243962.1 | XP_006244024.1 | 3 | 476 | |||
XM_006243963.2 | XP_006244025.1 | 3 | 476 | |||
XM_008766630.1 | XP_008764852.1 | 4 | 525 | |||
XM_008766631.1 | XP_008764853.1 | 3 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
