siRNA Details
Gene Symbol: Slc25a38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030032.1 | NP_001025203.1 | 4 | 567 | ||
XM_006244089.2 | XP_006244151.1 | 4 | 2468 | |||
GenBank mRNA | BC099762.1 | - | 4 | 567 | 7930329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
