siRNA Details
Gene Symbol: Eif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106867.1 | NP_001100337.1 | 2 | 264 | ||
GenBank mRNA | BC169125.1 | - | 2 | 184 | 6919533 | |
FQ211778.1 | - | 2 | 267 | |||
FQ214080.1 | - | 2 | 277 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
