siRNA Details
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106868.1 | NP_001100338.1 | 4, 5 | 458 | ||
XM_006244090.2 | XP_006244152.1 | 4, 5 | 1360 | |||
XM_006244091.2 | XP_006244153.1 | 4, 5 | 539 |
