siRNA Details
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106869.1 | NP_001100339.1 | 4 | 789 | ||
XM_006244189.2 | XP_006244251.1 | 4 | 780 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106869.1 | NP_001100339.1 | 4 | 789 | ||
XM_006244189.2 | XP_006244251.1 | 4 | 780 |